Potassium difluoroacetate 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC408095-1G
PC408095-5G
APOSPC408095-1GEA
147
EUR
APOSPC408095-1G
APOSPC408095-5G
Potassium difluoroacetate 95%
Potassium difluoroacetate
Formel:
C₂HF₂O₂·K Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD16620582 CAS-Nummer: 6291-26-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...