1-(2-Fluorobenzyl)hydrazine dihydrochloride
Lieferant: Apollo Scientific
Synonyme:
(2-Fluorobenzyl)hydrazine dihydrochloride
PC403063-100G
PC403063-250G
PC403063-50G
APOSPC403063-100GEA
424
EUR
APOSPC403063-100G
APOSPC403063-250G
APOSPC403063-50G
1-(2-Fluorobenzyl)hydrazine dihydrochloride
1-(2-Fluorobenzyl)hydrazine dihydrochloride
Formel:
C₇H₉FN₂·2ClH Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD17015850 CAS-Nummer: 1349715-77-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...