7-Bromo-4-(trifluoromethyl)quinolin-2(1H)-one 97%
Lieferant: Apollo Scientific
PC402130-1G
PC402130-500MG
APOSPC402130-1GEA
1720
EUR
APOSPC402130-1G
APOSPC402130-500MG
7-Bromo-4-(trifluoromethyl)quinolin-2(1H)-one 97%
7-Bromo-4-(trifluoromethyl)quinolin-2(1H)-one
Formel:
C₁₀H₅BrF₃NO Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD12756520 CAS-Nummer: 847900-73-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...