3-(4-Fluorophenyl)piperidine 95%

Lieferant: Apollo Scientific
Achtung

PC402101-1G PC402101-250MG
APOSPC402101-1GEA 342 EUR
APOSPC402101-1G APOSPC402101-250MG
3-(4-Fluorophenyl)piperidine 95%
3-(4-Fluorophenyl)piperidine
Formel: C₁₁H₁₄FN
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD08687917
CAS-Nummer: 676495-94-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR