5-Bromo-2-fluoro-3-methylbenzonitrile 98%

Lieferant: Apollo Scientific

PC402052-1G PC402052-5G
APOSPC402052-1GEA 358 EUR
APOSPC402052-1G APOSPC402052-5G
5-Bromo-2-fluoro-3-methylbenzonitrile 98%
5-Bromo-2-fluoro-3-methylbenzonitrile
Formel: C₈H₅BrFN
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD25955533
CAS-Nummer: 1110502-49-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR