Potassium trifluoro(oxetan-3-yl)borate

Lieferant: Apollo Scientific
Achtung

PC401500-100MG PC401500-1G PC401500-250MG
APOSPC401500-100MGEA 155 EUR
APOSPC401500-100MG APOSPC401500-1G APOSPC401500-250MG
Potassium trifluoro(oxetan-3-yl)borate
Potassium trifluoro(oxetan-3-yl)borate
Formel: C₃H₅BF₃O·K
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD24843091
CAS-Nummer: 1430219-76-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR