tert-Butyl 2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]acetate

Lieferant: Apollo Scientific

PC401032-1G PC401032-5G
APOSPC401032-1GEA 657 EUR
APOSPC401032-1G APOSPC401032-5G
tert-Butyl 2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]acetate
tert-Butyl 2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]acetate
Formel: C₁₂H₁₃ClF₃NO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD28122853
CAS-Nummer: 1672655-80-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR