2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]acetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(3-Chloro-5-(trifluoromethyl)pyridin-2-yl)acetic acid

PC401029-1G PC401029-250MG
APOSPC401029-1GEA 265 EUR
APOSPC401029-1G APOSPC401029-250MG
2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]acetic acid
2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]acetic acid
Formel: C₈H₅ClF₃NO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09925056
CAS-Nummer: 1000522-34-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR