2-(Trifluoromethyl)pyrimidine-4,6-diamine

Lieferant: Apollo Scientific

Synonyme: 4,6-Diamino-2-(trifluoromethyl)pyrimidine, 4,6-Diamino-2-(trifluoromethyl)-1,3-diazine

PC400533-1G PC400533-250MG
APOSPC400533-1GEA 321 EUR
APOSPC400533-1G APOSPC400533-250MG
2-(Trifluoromethyl)pyrimidine-4,6-diamine
2-(Trifluoromethyl)pyrimidine-4,6-diamine
Formel: C₅H₅F₃N₄
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09743717
CAS-Nummer: 672-46-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR