3-Fluoropiperidine-3-carbonitrile 95%
Lieferant: Apollo Scientific
PC400522-250MG
PC400522-500MG
APOSPC400522-250MGEA
567
EUR
APOSPC400522-250MG
APOSPC400522-500MG
3-Fluoropiperidine-3-carbonitrile 95%
3-Fluoropiperidine-3-carbonitrile
Formel:
C₆H₉FN₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD22690799 CAS-Nummer: 1265323-71-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...