1-(5-Bromo-4-methylpyridin-2-yl)-2,2,2-trifluoroethanone

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(5-Bromo-4-methylpyridin-2-yl)-2,2,2-trifluoroethan-1-one

PC400149-1G PC400149-250MG
APOSPC400149-1GEA 556 EUR
APOSPC400149-1G APOSPC400149-250MG
1-(5-Bromo-4-methylpyridin-2-yl)-2,2,2-trifluoroethanone
1-(5-Bromo-4-methylpyridin-2-yl)-2,2,2-trifluoroethanone
Formel: C₈H₅BrF₃NO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD31621154
CAS-Nummer: 2231676-74-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR