3-Bromo-2-fluoro-6-nitrotoluene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-2-fluoro-3-methyl-4-nitrobenzene, 4-Bromo-3-fluoro-2-methylnitrobenzene
PC400013-1G
PC400013-250MG
APOSPC400013-1GEA
209
EUR
APOSPC400013-1G
APOSPC400013-250MG
3-Bromo-2-fluoro-6-nitrotoluene
3-Bromo-2-fluoro-6-nitrotoluene
Formel:
C₇H₅BrFNO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD26407497 CAS-Nummer: 1807209-28-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...