3,4-Dichloro-2-fluorobenzenesulphonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
PC37866-1G
PC37866-250MG
PC37866-5G
APOSPC37866-1GEA
575
EUR
APOSPC37866-1G
APOSPC37866-250MG
APOSPC37866-5G
3,4-Dichloro-2-fluorobenzenesulphonyl chloride
3,4-Dichloro-2-fluorobenzenesulphonyl chloride
Formel:
C₆H₂Cl₃FO₂S Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD28785980 CAS-Nummer: 1803718-50-4 UN-Nummer: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...