2-Amino-4-bromo-5-fluorophenol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-4-fluoro-2-hydroxyaniline
PC3691-1G
PC3691-250MG
PC3691-5G
APOSPC3691-1GEA
151
EUR
APOSPC3691-1G
APOSPC3691-250MG
APOSPC3691-5G
2-Amino-4-bromo-5-fluorophenol
2-Amino-4-bromo-5-fluorophenol
Formel:
C₆H₅BrFNO Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD11101429 CAS-Nummer: 1016234-89-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...