2-Bromo-5-fluoro-1-cyclopropylbenzene 95%

Lieferant: Apollo Scientific
Achtung

PC32945-1G PC32945-250MG PC32945-5G
APOSPC32945-1GEA 347 EUR
APOSPC32945-1G APOSPC32945-250MG APOSPC32945-5G
2-Bromo-5-fluoro-1-cyclopropylbenzene 95%
2-Bromo-5-fluoro-1-cyclopropylbenzene
Formel: C₉H₈BrF
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD21100002
CAS-Nummer: 1353853-80-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR