2-[Bromo(difluoro)methyl]-4-chloro-1H-benzimidazole

Lieferant: Apollo Scientific
Achtung

PC32921-1G PC32921-250MG PC32921-5G
APOSPC32921-1GEA 53.8 EUR
APOSPC32921-1G APOSPC32921-250MG APOSPC32921-5G
2-[Bromo(difluoro)methyl]-4-chloro-1H-benzimidazole
2-[Bromo(difluoro)methyl]-4-chloro-1H-benzimidazole
Formel: C₈H₄BrClF₂N₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD31922504
CAS-Nummer: 2366994-65-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR