2-[Bromo(difluoro)methyl]-4-fluoro-1H-benzimidazole
Lieferant: Apollo Scientific
|
Achtung
|
PC32919-1G
PC32919-250MG
PC32919-5G
APOSPC32919-1GEA
52.2
EUR
APOSPC32919-1G
APOSPC32919-250MG
APOSPC32919-5G
2-[Bromo(difluoro)methyl]-4-fluoro-1H-benzimidazole
2-[Bromo(difluoro)methyl]-4-fluoro-1H-benzimidazole
Formel:
C₈H₄BrF₃N₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD31922502 CAS-Nummer: 2366994-67-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...