5-Bromo-3-fluoro-2-nitrophenol

Lieferant: Apollo Scientific
Achtung

PC32840-1G PC32840-250MG PC32840-5G
APOSPC32840-1GEA 200 EUR
APOSPC32840-1G APOSPC32840-250MG APOSPC32840-5G
5-Bromo-3-fluoro-2-nitrophenol
5-Bromo-3-fluoro-2-nitrophenol
Formel: C₆H₃BrFNO₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD28348375
CAS-Nummer: 1807256-50-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR