3-Fluoro-5-formylbenzeneboronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Fluoro-5-formylphenyl)boronic acid, 3-Borono-5-fluorobenzaldehyde, 3-Fluoro-5-formylphenylboronic acid

PC32759-1G PC32759-5G PC32759-250MG
APOSPC32759-1GEA 29 EUR
APOSPC32759-1G APOSPC32759-5G APOSPC32759-250MG
3-Fluoro-5-formylbenzeneboronic acid
3-Fluoro-5-formylbenzeneboronic acid
Formel: C₇H₆BFO₃
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD11856017
CAS-Nummer: 328956-60-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR