Pentafluorophenyl 4-(3,5-dimethyl-1H-pyrazol-1-yl)benzoate 97%

Lieferant: Apollo Scientific

PC3226-1G PC3226-250MG
APOSPC3226-1GEA 220 EUR
APOSPC3226-1G APOSPC3226-250MG
Pentafluorophenyl 4-(3,5-dimethyl-1H-pyrazol-1-yl)benzoate 97%
Pentafluorophenyl 4-(3,5-dimethyl-1H-pyrazol-1-yl)benzoate
Formel: C₁₈H₁₁F₅N₂O₂
Schmelzpunkt: 85.5…86 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09879978
CAS-Nummer: 941717-00-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR