2,4-Difluoro-N-{2-[(fur-2-ylmethyl)thio]ethyl}benzenesulphonamide
Lieferant: Apollo Scientific
PC31871-1G
PC31871-250MG
APOSPC31871-1GEA
102
EUR
APOSPC31871-1G
APOSPC31871-250MG
2,4-Difluoro-N-{2-[(fur-2-ylmethyl)thio]ethyl}benzenesulphonamide
2,4-Difluoro-N-{2-[(fur-2-ylmethyl)thio]ethyl}benzenesulphonamide
Formel:
C₁₃H₁₃F₂NO₃S₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01570573 CAS-Nummer: 680211-96-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...