methyl 2-(5-fluoro-2-nitrophenoxy)acetate
Lieferant: Apollo Scientific
PC31301-1G
PC31301-250MG
PC31301-500MG
PC31301-2.5G
APOSPC31301-1GEA
139
EUR
APOSPC31301-1G
APOSPC31301-250MG
APOSPC31301-500MG
APOSPC31301-2.5G
methyl 2-(5-fluoro-2-nitrophenoxy)acetate
methyl 2-(5-fluoro-2-nitrophenoxy)acetate
Formel:
C₉H₈FNO₅ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00117826 CAS-Nummer: 116355-65-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...