3-Bromo-2-chloro-5-(trifluoromethyl)benzoyl chloride
Lieferant: Apollo Scientific
PC303582-1G
PC303582-5G
APOSPC303582-1GEA
231
EUR
APOSPC303582-1G
APOSPC303582-5G
3-Bromo-2-chloro-5-(trifluoromethyl)benzoyl chloride
3-Bromo-2-chloro-5-(trifluoromethyl)benzoyl chloride
Formel:
C₈H₂BrCl₂F₃O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD32640159 CAS-Nummer: 2169633-17-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...