3-Bromo-2-chloro-5-(trifluoromethyl)benzyl bromide 97%
Lieferant: Apollo Scientific
PC303577-1G
PC303577-5G
APOSPC303577-1GEA
279
EUR
APOSPC303577-1G
APOSPC303577-5G
3-Bromo-2-chloro-5-(trifluoromethyl)benzyl bromide 97%
3-Bromo-2-chloro-5-(trifluoromethyl)benzyl bromide
Formel:
C₈H₄Br₂ClF₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD32640154 CAS-Nummer: 2091543-20-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...