3-Bromo-4-chloro-5-(trifluoromethyl)phenylacetic acid

Lieferant: Apollo Scientific

PC303567-1G PC303567-5G
APOSPC303567-1GEA 418 EUR
APOSPC303567-1G APOSPC303567-5G
3-Bromo-4-chloro-5-(trifluoromethyl)phenylacetic acid
3-Bromo-4-chloro-5-(trifluoromethyl)phenylacetic acid
Formel: C₉H₅BrClF₃O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD32640145
CAS-Nummer: 2386381-95-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR