2-Bromo-5-fluoro-4-(trifluoromethyl)benzamide
Lieferant: Apollo Scientific
PC303559-1G
PC303559-5G
APOSPC303559-1GEA
290
EUR
APOSPC303559-1G
APOSPC303559-5G
2-Bromo-5-fluoro-4-(trifluoromethyl)benzamide
2-Bromo-5-fluoro-4-(trifluoromethyl)benzamide
Formel:
C₈H₄BrF₄NO Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD32640137 CAS-Nummer: 2090465-20-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...