3-Bromo-6-fluoro-2-(trifluoromethyl)benzyl alcohol 97%

Lieferant: Apollo Scientific

PC303535-1G PC303535-5G
APOSPC303535-1GEA 231 EUR
APOSPC303535-1G APOSPC303535-5G
3-Bromo-6-fluoro-2-(trifluoromethyl)benzyl alcohol 97%
3-Bromo-6-fluoro-2-(trifluoromethyl)benzyl alcohol
Formel: C₈H₅BrF₄O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD32640117
CAS-Nummer: 2090282-94-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR