5-Bromo-4-chloro-2-fluorobenzonitrile

Lieferant: Apollo Scientific

PC303484-1G PC303484-5G
APOSPC303484-1GEA 325 EUR
APOSPC303484-1G APOSPC303484-5G
5-Bromo-4-chloro-2-fluorobenzonitrile
5-Bromo-4-chloro-2-fluorobenzonitrile
Formel: C₇H₂BrClFN
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD20486525
CAS-Nummer: 1824089-00-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR