5-Bromo-3-chloro-2-fluorobenzyl bromide 95%

Lieferant: Apollo Scientific

PC303454-1G PC303454-250MG
APOSPC303454-1GEA 667 EUR
APOSPC303454-1G APOSPC303454-250MG
5-Bromo-3-chloro-2-fluorobenzyl bromide 95%
5-Bromo-3-chloro-2-fluorobenzyl bromide
Formel: C₇H₄Br₂ClF
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD32640076
CAS-Nummer: 2092617-82-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR