6-Chloro-2,4-difluoro-3-methylbenzamide 97%
Lieferant: Apollo Scientific
PC303405-1G
PC303405-250MG
APOSPC303405-1GEA
927
EUR
APOSPC303405-1G
APOSPC303405-250MG
6-Chloro-2,4-difluoro-3-methylbenzamide 97%
6-Chloro-2,4-difluoro-3-methylbenzamide
Formel:
C₈H₆ClF₂NO Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD32640059 CAS-Nummer: 2415751-80-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...