3,4-Dichloro-5-(trifluoromethyl)phenylacetic acid 97%
Lieferant: Apollo Scientific
PC303242-1G
PC303242-250MG
APOSPC303242-1GEA
990
EUR
APOSPC303242-1G
APOSPC303242-250MG
3,4-Dichloro-5-(trifluoromethyl)phenylacetic acid 97%
3,4-Dichloro-5-(trifluoromethyl)phenylacetic acid
Formel:
C₉H₅Cl₂F₃O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD28103374 CAS-Nummer: 1823541-32-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...