2-Fluoro-4-methyl-3-(trifluoromethyl)benzoic acid 97%

Lieferant: Apollo Scientific

PC303155-1G PC303155-250MG
APOSPC303155-1GEA 507 EUR
APOSPC303155-1G APOSPC303155-250MG
2-Fluoro-4-methyl-3-(trifluoromethyl)benzoic acid 97%
2-Fluoro-4-methyl-3-(trifluoromethyl)benzoic acid
Formel: C₉H₆F₄O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD28103359
CAS-Nummer: 1824048-54-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR