4-Chloro-2-(trifluoromethyl)cinnamic acid 98%
Lieferant: Apollo Scientific
Synonyme:
(E)-3-(4-chloro-2-(trifluoromethyl)phenyl)acrylic acid
PC302970-10G
PC302970-5G
PC302970-1G
APOSPC302970-10GEA
405
EUR
APOSPC302970-10G
APOSPC302970-5G
APOSPC302970-1G
4-Chloro-2-(trifluoromethyl)cinnamic acid 98%
4-Chloro-2-(trifluoromethyl)cinnamic acid
Formel:
C₁₀H₆ClF₃O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06205746 CAS-Nummer: 773132-29-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...