Ethyl 6-fluoro-1H-indole-2-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Ethoxycarbonyl)-6-fluoro-1H-indole

PC3029-1G PC3029-5G
APOSPC3029-1GEA 34.8 EUR
APOSPC3029-1G APOSPC3029-5G
Ethyl 6-fluoro-1H-indole-2-carboxylate
Ethyl 6-fluoro-1H-indole-2-carboxylate
Formel: C₁₁H₁₀FNO₂
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD03095203
CAS-Nummer: 348-37-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR