4-Chloro-3-fluoro-DL-phenylglycine

Lieferant: Apollo Scientific
Achtung

PC302581-1G PC302581-5G
APOSPC302581-1GEA 230 EUR
APOSPC302581-1G APOSPC302581-5G
4-Chloro-3-fluoro-DL-phenylglycine
4-Chloro-3-fluoro-DL-phenylglycine
Formel: C₈H₇ClFNO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD04116049
CAS-Nummer: 1039338-62-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR