4-(Trifluoromethoxy)-DL-phenylalanine
Lieferant: Apollo Scientific
|
Achtung
|
PC302564-1G
PC302564-5G
PC302564-10G
APOSPC302564-1GEA
109
EUR
APOSPC302564-1G
APOSPC302564-5G
APOSPC302564-10G
4-(Trifluoromethoxy)-DL-phenylalanine
4-(Trifluoromethoxy)-DL-phenylalanine
Formel:
C₁₀H₁₀F₃NO₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00764461 CAS-Nummer: 174732-57-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...