3-Chloro-2-fluoro-DL-phenylglycine
Lieferant: Apollo Scientific
PC302522-1G
PC302522-5G
APOSPC302522-1GEA
159
EUR
APOSPC302522-1G
APOSPC302522-5G
3-Chloro-2-fluoro-DL-phenylglycine
3-Chloro-2-fluoro-DL-phenylglycine
Formel:
C₈H₇ClFNO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD04116016 CAS-Nummer: 1042665-36-0 UN-Nummer: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...