2-Fluoro-5-methyl-DL-phenylglycine

Lieferant: Apollo Scientific

PC302461-1G PC302461-5G PC302461-10G
APOSPC302461-1GEA 174 EUR
APOSPC302461-1G APOSPC302461-5G APOSPC302461-10G
2-Fluoro-5-methyl-DL-phenylglycine
2-Fluoro-5-methyl-DL-phenylglycine
Formel: C₉H₁₀FNO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06660229
CAS-Nummer: 1260004-37-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR