2,3,4-Trifluoro-DL-phenylalanine

Lieferant: Apollo Scientific

PC302423-1G PC302423-5G PC302423-10G
APOSPC302423-1GEA 111 EUR
APOSPC302423-1G APOSPC302423-5G APOSPC302423-10G
2,3,4-Trifluoro-DL-phenylalanine
2,3,4-Trifluoro-DL-phenylalanine
Formel: C₉H₈F₃NO₂
Molekulargewicht: 219.161 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD04116037
CAS-Nummer: 1259978-34-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR