4-Chloro-2-(trifluoromethyl)benzyl alcohol 98%

Lieferant: Apollo Scientific
Achtung

PC302408-5G PC302408-10G PC302408-1G
APOSPC302408-5GEA 143 EUR
APOSPC302408-5G APOSPC302408-10G APOSPC302408-1G
4-Chloro-2-(trifluoromethyl)benzyl alcohol 98%
4-Chloro-2-(trifluoromethyl)benzyl alcohol
Formel: C₈H₆ClF₃O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD04115839
CAS-Nummer: 773872-13-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR