4-Fluoro-3,5-dimethyl-DL-phenylalanine
Lieferant: Apollo Scientific
PC302150-1G
PC302150-5G
PC302150-10G
APOSPC302150-1GEA
116
EUR
APOSPC302150-1G
APOSPC302150-5G
APOSPC302150-10G
4-Fluoro-3,5-dimethyl-DL-phenylalanine
4-Fluoro-3,5-dimethyl-DL-phenylalanine
Formel:
C₁₁H₁₄FNO₂ Schmelzpunkt: 245 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09832378 CAS-Nummer: 1259957-45-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...