3-Fluoro-4-(trifluoromethyl)-DL-phenylalanine
Lieferant: Apollo Scientific
|
Achtung
|
PC302141-1G
PC302141-5G
PC302141-10G
APOSPC302141-1GEA
116
EUR
APOSPC302141-1G
APOSPC302141-5G
APOSPC302141-10G
3-Fluoro-4-(trifluoromethyl)-DL-phenylalanine
3-Fluoro-4-(trifluoromethyl)-DL-phenylalanine
Formel:
C₁₀H₉F₄NO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD04115998 CAS-Nummer: 1215491-31-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...