3-Chloro-4-fluoro-DL-phenylalanine

Lieferant: Apollo Scientific
Achtung

PC302139-1G PC302139-5G PC302139-10G
APOSPC302139-1GEA 116 EUR
APOSPC302139-1G APOSPC302139-5G APOSPC302139-10G
3-Chloro-4-fluoro-DL-phenylalanine
3-Chloro-4-fluoro-DL-phenylalanine
Formel: C₉H₉ClFNO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD04116027
CAS-Nummer: 7731-00-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR