2-Fluoro-3-(trifluoromethyl)-DL-phenylalanine

Lieferant: Apollo Scientific

PC302131-1G PC302131-5G PC302131-10G
APOSPC302131-1GEA 171 EUR
APOSPC302131-1G APOSPC302131-5G APOSPC302131-10G
2-Fluoro-3-(trifluoromethyl)-DL-phenylalanine
2-Fluoro-3-(trifluoromethyl)-DL-phenylalanine
Formel: C₁₀H₉F₄NO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD04116000
CAS-Nummer: 1259993-82-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR