2-Chloro-6-fluoro-DL-phenylalanine
Lieferant: Apollo Scientific
PC302130-1G
PC302130-5G
PC302130-10G
APOSPC302130-1GEA
116
EUR
APOSPC302130-1G
APOSPC302130-5G
APOSPC302130-10G
2-Chloro-6-fluoro-DL-phenylalanine
2-Chloro-6-fluoro-DL-phenylalanine
Formel:
C₉H₉ClFNO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD04116005 CAS-Nummer: 603940-86-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...