2-Chloro-5-(trifluoromethyl)-DL-phenylalanine

Lieferant: Apollo Scientific

PC302128-1G PC302128-5G PC302128-10G
APOSPC302128-1GEA 116 EUR
APOSPC302128-1G APOSPC302128-5G APOSPC302128-10G
2-Chloro-5-(trifluoromethyl)-DL-phenylalanine
2-Chloro-5-(trifluoromethyl)-DL-phenylalanine
Formel: C₁₀H₉ClF₃NO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06660185
CAS-Nummer: 64134-20-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR