2,3-Dichloro-6-fluoro-DL-phenylalanine
Lieferant: Apollo Scientific
PC302120-1G
PC302120-5G
PC302120-10G
PC302120-25G
APOSPC302120-1GEA
159
EUR
APOSPC302120-1G
APOSPC302120-5G
APOSPC302120-10G
APOSPC302120-25G
2,3-Dichloro-6-fluoro-DL-phenylalanine
2,3-Dichloro-6-fluoro-DL-phenylalanine
Formel:
C₉H₈Cl₂FNO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06660177 CAS-Nummer: 1259973-77-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...