5-Methyl-2-(trifluoromethyl)benzyl alcohol 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC302119-1G
PC302119-5G
APOSPC302119-1GEA
69.6
EUR
APOSPC302119-1G
APOSPC302119-5G
5-Methyl-2-(trifluoromethyl)benzyl alcohol 98%
5-Methyl-2-(trifluoromethyl)benzyl alcohol
Formel:
C₉H₉F₃O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06660295 CAS-Nummer: 886502-81-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...