1-(2,4-Difluoro-6-hydroxyphenyl)ethanone
Lieferant: Apollo Scientific
|
Achtung
|
PC301028-1G
PC301028-250MG
APOSPC301028-1GEA
986
EUR
APOSPC301028-1G
APOSPC301028-250MG
1-(2,4-Difluoro-6-hydroxyphenyl)ethanone
1-(2,4-Difluoro-6-hydroxyphenyl)ethanone
Formel:
C₈H₆F₂O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD18910100 CAS-Nummer: 1356999-25-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...