2-Methyl-3-(1H-pyrazol-5-yl)-6-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific
Achtung

PC300770-1G PC300770-100MG PC300770-250MG
APOSPC300770-1GEA 139 EUR
APOSPC300770-1G APOSPC300770-100MG APOSPC300770-250MG
2-Methyl-3-(1H-pyrazol-5-yl)-6-(trifluoromethyl)pyridine
2-Methyl-3-(1H-pyrazol-5-yl)-6-(trifluoromethyl)pyridine
Formel: C₁₀H₈F₃N₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD27995624
CAS-Nummer: 1818100-16-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR